Learn More
3M™ Harvest RC Centrate Chromatographic Clarifier, BC25, Luer
Description
- Single-stage centrate clarification for process compression and streamlining of process development.
- Enables de-risking of clarification process development, scale-up and tech transfer for faster speed to market.
- Synthetic, functionalized fibrous media provides effective soluble impurity removal in centrate, including DNA and cell shear-related submicron particle reduction.
- Predictable performance scalability for consistent centrate clarification at various process scales.
- The single-mode electrostatic separation mechanism and high-throughput media enable product recoveries of >95%.
- Provides comparable effluent quality to 3M™ Harvest RC Chromatographic Clarifier for a modern clarification platform.
- Lower consumption of buffer or water compared to depth filtration.
- Alkaline-resistant capsules for pre-use sanitization.
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Filter Capsule |
| Material (Membrane) | Polyamide |
| Sterility | Non-sterile |
| Quantity | 2/Cs. |
| Type | Chromatography Filter |
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?