missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Needle L-Shaped Platinum Electrode Kit for Electroporators
Click to view available options
Pulse Form:
Square Wave
Unit Size:
Each
Description
- Accessory for Electroporators
- Enables pinpoint transfection for greater ease and efficiency in fragile tissue
- L-shaped electrodes available in 3 mm tip length
Specifications
Specifications
| Format | Kit |
| For Use With (Application) | Ex-vivo or in-vivo tissues gene or drug delivery. Nuclear Transfer |
| For Use With (Equipment) | ECM830, Gemini X2, ECM2001+ |
| Material | Platinum |
| Product Line | BTX Electroporation |
| Product Type | L-shaped Electrodes |
| Voltage | 100 V |
| Electrode Shape | L-shaped |
| Includes | 2 extended L-shaped platinum needles ultra thin electrodes and connecting cable |
| Length (English) Electrode | 0.12 in. |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction