missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | HARS2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18323434
|
Novus Biologicals
NBP3-04858-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373437
|
Novus Biologicals
NBP3-04858-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HARS2 Polyclonal specifically detects HARS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HARS2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 23438 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EC 6.1.1, EC 6.1.1.21, HARSLprobable histidyl-tRNA synthetase, mitochondrial, HARS-related, HARSRhistidine tRNA ligase 2, mitochondrial (putative), HisRS, histidine translase, Histidine--tRNA ligase, Histidine--tRNA ligase-like, histidyl-tRNA synthetase 2, mitochondrial (putative), histidyl-tRNA synthetase-like, HO3histidine-tRNA ligase homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 217-506 of human HARS2 (NP_036340.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title