missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HARS Recombinant Rabbit Monoclonal Antibody (K01_1S03)
GREENER_CHOICE

Product Code. 30828609
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828609 100 μL
30829575 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828609 Supplier Invitrogen™ Supplier No. MA563646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HARS Recombinant Monoclonal Antibody for Western Blot

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HARS
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1S03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene HARS1
Gene Accession No. P12081
Gene Alias CMT2W; dead end homolog 1; Dnd1; FLJ20491; HARS; HARS1; HARSL; HARSL protein; HisRS; histidine translase; histidine tRNA ligase; histidine--tRNA ligase; histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 1; histidyl-tRNA synthetase, cytoplasmic; HRS; im:7136719; MMHRS; Unknown (protein for IMAGE:7951312); USH3B; zgc:92215
Gene Symbols HARS1
Host Species Rabbit
Immunogen A synthetic peptide of human HARS.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3035
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.