missing translation for 'onlineSavingsMsg'
Learn More

Harringtonine, MedChemExpress

Product Code. 16429148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429148 10 mg 10mg
16481548 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16429148 Supplier Medchem Express Supplier No. HYN0862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Harringtonine is a natural Cephalotaxus alkaloid that inhibits protein synthesis. Harringtonine has anti-chikungunya virus (CHIKV) activities with an EC50 of 0.24 μM.

Harringtonine is a natural Cephalotaxus alkaloid that inhibits protein synthesis. Harringtonine has anti-chikungunya virus (CHIKV) activities with an EC50 of 0.24 μM.

Specifications

Chemical Name or Material Harringtonine
CAS 26833-85-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H37NO9
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (188.11 mM; Need ultrasonic)
SMILES O=C([C@@](CC(OC)=O)(O)CCC(C)(O)C)O[C@H]1[C@](C2=CC(OCO3)=C3C=C2CCN4CCC5)([H])[C@]45C=C1OC
Molecular Weight (g/mol) 531.59
Formula Weight 531.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.