missing translation for 'onlineSavingsMsg'
Learn More

Harmine, 98%

Product Code. 10412071 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10412071

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 442-51-3 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Melting Point 262.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 4649
Solubility Information Solubility in water: insoluble.
Formula Weight 212.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Harmine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Harmine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Harmine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.