missing translation for 'onlineSavingsMsg'
Learn More

Harmine, 98+%

Product Code. p-7047382
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11359615

Brand: Thermo Scientific Alfa Aesar L19068.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 442-51-3 | C13H12N2O | 212.25 g/mol

Harmine, inhibits the Wnt signaling pathway in a cell-specific manner. Down-regulates inflammatory gene expression (TNF-α, IL-1β, iNOS). Reversibly inhibits MAO-A (monoamine oxidase A) but has no effect on MAO-B. Harmine is an inhibitor of Dyrk1A, Dyrk2 and Dyrk3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Harmine, inhibits the Wnt signaling pathway in a cell-specific manner. Down-regulates inflammatory gene expression (TNF-α, IL-1β, iNOS). Reversibly inhibits MAO-A (monoamine oxidase A) but has no effect on MAO-B. Harmine is an inhibitor of Dyrk1A, Dyrk2 and Dyrk3.

Solubility
Soluble in DMSO (100 mM), DMF (∼1.5 mg/ml), ethanol (∼1.5 mg/ml), and PBS (pH 7.2, ∼0.25 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Melting Point 262°C to 266°C
Odor Odorless
Quantity 250 mg
Beilstein 178813
Merck Index 14,4616
Solubility Information Soluble in DMSO (100 mM),DMF (∽1.5mg/ml),ethanol (∽1.5mg/ml),and PBS (pH 7.2,∽0.25mg/ml). Insoluble in water.
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material Harmine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.