Learn More
Lovibond™ HARDCHECK P Total Hardness Tablets
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 100
Description
The suitable tablet for the fast result
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Specifications
| Quantity | 100 Tablets |
| Range | 2 to 50 mg/L CaCO3 and 20 to 500 mg/L CaCO3 |
| Type | Reagent |