Learn More
Faster™ Hard Exhaust Duct Connection for SafeFAST and CytoFAST Cabinets
Product Code. 16268590
Click to view available options
For Use With (Equipment):
SafeFAST, CytoFAST 109/215/315
SafeFAST, CytoFAST 112/115/218/318
SafeFAST, CytoFAST 209
SafeFAST, CytoFAST 212/312
Product Code. 16268590
Brand: Faster™ FX0002154040
Hard duct connection on the top of the cabinet to connect to ductwork.
  • 200 mm diameter
  • Specifically for SafeFAST or CytoFAST cabinets
Specifications
For Use With (Application) Ducting Safety Cabinets
For Use With (Equipment) SafeFAST, CytoFAST 109/215/315