Learn More
Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop
Use Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop for general laboratory needs including plating and quick and accurate sample throughput.
Brand:  Technical Service Consultants™ TS/2-C
Code : 03
Additional Details : Weight : 2.45700kg
 View more versions of this product
Product Code. 12799865
  • £550.00 / Pack of 1000
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Cadmium-free pigments
  • Gamma Irradiation to achieve a SAL of 10-6
  • Shelf Life: 3 years
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Specifications
Yes
Hard Crystal Polystyrene
Green
1000/Pk.
Sterile
10 μL
Inoculating Loop
Singles in peel pouch