Learn More
Invitrogen™ Haptoglobin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522255
Description
Recommended positive controls: HeLa, HepG2, A375, human plasma. Predicted reactivity: Mouse (83%), Rat (80%), Cat (84%), Pig (82%), Rabbit (87%), Rhesus Monkey (96%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.
Specifications
| Haptoglobin | |
| Polyclonal | |
| Unconjugated | |
| HP | |
| alpha polypeptide; Ba1-647; beta polypeptide; binding peptide; Binding peptide (BP); BP; haptoglobin; haptoglobin alpha 1S; Haptoglobin alpha chain; Haptoglobin alpha chain / alpha(1S) beta; haptoglobin alpha(1S)-beta; Haptoglobin alpha(2FS) beta; haptoglobin alpha(2FS)-beta; Haptoglobin beta chain; haptoglobin, alpha polypeptide; haptoglobin, beta polypeptide; haptoglobin; haptoglobin-like; haptoglobin-like; haptoglobin-related protein; Hp; HP-1; HP2 ALPHA2; HP2ALPHA2; HP2-ALPHA-2; HPA1S; HPR; HPT; Liver regeneration-related protein LRRG173; LOC100067869; LOC479668; MGC111141; MGC133752; preHP2; zonulin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P00738 | |
| HP | |
| Recombinant protein encompassing a sequence within the center region of human Haptoglobin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.