missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32391
This item is not returnable.
View return policy
Description
Haptoglobin Polyclonal specifically detects Haptoglobin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Haptoglobin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.01% Thimerosal | |
| HP | |
| Recombinant protein encompassing a sequence within the center region of human Haptoglobin. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 3240 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.59mg/mL | |
| Western Blot 1:5000-1:30000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction