missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haptoglobin Antibody (5A5), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Haptoglobin |
|---|---|
| Clone | 5A5 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031113
|
Novus Biologicals
NBP3-26220-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30025659
|
Novus Biologicals
NBP3-26220-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Haptoglobin | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| A synthesized peptide derived from Human Haptoglobin [UniProt P00738] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5A5 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3240 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title