missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP), Rat anti-Non-species specific, Clone: B1-8, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-81028-0.025MG
This item is not returnable.
View return policy
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control.
Specifications
| hapten 4-hydroxy-3-nitrophenyl acetyl (NP) | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Control | |
| Rat | |
| Protein A purified | |
| RUO | |
| Hapten (4-hydroxy-3-nitropheny1) acetyl (NP). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 λ |
| Western Blot, Flow Cytometry | |
| B1-8 | |
| Unconjugated | |
| PBS | |
| hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
| 0.025 mg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction