missing translation for 'onlineSavingsMsg'
Learn More

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8), HRP, Novus Biologicals™

Product Code. 18318980 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318980 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318980 Supplier Novus Biologicals Supplier No. NBP281027H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hapten 4-hydroxy-3-nitrophenyl acetyl (NP)
Applications Western Blot
Classification Monoclonal
Clone B1-8
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias (4-Hydroxy-3-nitrophenyl)acetyl, 2-(4-Hydroxy-3-nitrophenyl)acetate, 2-(4-Hydroxy-3-nitrophenyl)acetic acid, 3-nitro-4-Hydroxyphenylacetic acid, 4-Hydroxy-3-nitrophenyl acetyl
Host Species Human
Immunogen hapten-protein conjugated NP12-chicken gammaglobulin (CG).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.