missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52692-0.025MG
This item is not returnable.
View return policy
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF-ready.
Specifications
| hapten 4-hydroxy-3-nitrophenyl acetyl (NP) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
| 0.025 mg | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| B1-8 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Hapten (4-hydroxy-3-nitropheny1) acetyl (NP). | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction