missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAPLN4 Monoclonal Antibody (387915)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561383
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Hyaluronan and proteoglycan link protein 4 (HAPLN4), also known as Brain Link Protein 4 (BRAL1), is a gene located on chromosome 5q14.2, encoding a member of the link protein family that plays a crucial role in stabilizing interactions between hyaluronan and proteoglycans within the extracellular matrix. HAPLN4 is predominantly expressed in the brain, especially within the central nervous system where it contributes to the structural integrity and functional modulation of neural tissues. The protein is key in maintaining the organization of the extracellular matrix by binding hyaluronan and other matrix components, facilitating cell adhesion and signaling. This is vital for processes like neuronal migration, synapse formation, and neuronal differentiation during brain development. Variations in HAPLN4 expression or function are associated with neural developmental disorders and may influence the progression of neurodegenerative diseases due to its critical role in preserving the extracellular matrix architecture and neural connectivity.
Specifications
| HAPLN4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86UW8 | |
| HAPLN4 | |
| Mouse myeloma cell line NS0-derived recombinant human HAPLN4, Gln30-Val402 (predicted). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 387915 | |
| Unconjugated | |
| HAPLN4 | |
| 9330174O11; Brain link protein 2; brain linked protein 2; Bral2; Hapln4; Hyaluronan and proteoglycan link protein 4; hyaluronan and proteoglycan link protein 4; LOW QUALITY PROTEIN: hyaluronan and proteoglycan link protein 4; KIAA1926; link protein 4; LOW QUALITY PROTEIN: hyaluronan and proteoglycan link protein 4; Lpr4; si:dkey-19f23.2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 404037 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction