missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | HAPLN4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HAPLN4 Polyclonal specifically detects HAPLN4 in Human samples. It is validated for Western Blot.Specifications
| HAPLN4 | |
| Polyclonal | |
| Rabbit | |
| NP_075378 | |
| 404037 | |
| Synthetic peptide directed towards the C terminal of human HAPLN4The immunogen for this antibody is HAPLN4. Peptide sequence NYGYRHNAEERYDAFCFTSNLPGRVFFLKPLRPVPFSGAARACAARGAAV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BRAL2KIAA1926Brain link protein 2, hyaluronan and proteoglycan link protein 4 | |
| HAPLN4 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title