missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77322PCP
This item is not returnable.
View return policy
Description
HAPLN2 Polyclonal antibody specifically detects HAPLN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HAPLN2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| Brain link protein 1, brain link protein-1, BRAL1, hyaluronan and proteoglycan link protein 2 | |
| Antibody was raised against a 12 amino acid synthetic peptide from near the center of human BRAL1. The immunogen is located within amino acids 130 - 180 of BRAL1. Amino Acid Squence: FPYQPSRGRYQF | |
| 0.1 mL | |
| Neuroscience | |
| 60484 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction