missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN1 Recombinant Rabbit Monoclonal Antibody (5O9U2)

HAPLN1 Recombinant Rabbit Monoclonal Antibody (ARC0234)
Brand: Invitrogen MA535583
This item is not returnable.
View return policy
Description
HAPLN1 Recombinant Monoclonal Antibody for Western Blot
Specifications
| HAPLN1 | |
| Recombinant Monoclonal | |
| 2.27 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P03994, P10915, Q9QUP5 | |
| HAPLN1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-354 of human HAPLN1 (P10915). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5O9U2 | |
| Unconjugated | |
| HAPLN1 | |
| BB099155; Cartilage link protein; cartilage link protein 1; cartilage linking protein 1; cartilage-link protein; Cartilage-linking protein 1; CLP; CRT1; Crtl1; Crtl1l; fc26f11; fc37g04; fc50e12; fc55h01; HAPLN1; hapln1a; Hyaluronan and proteoglycan link protein 1; hyaluronan and proteoglycan link protein 1a; hyaluronan proteoglycan link protein 1; link protein; LP; LP-1; Proteoglycan link protein; wu:fc26f11; wu:fc37g04; wu:fc50e12; wu:fc55h01 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12950, 1404, 29331 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction