Learn More
Invitrogen™ HAPLN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593098
Description
Immunogen sequence: PRLGRYNLNF HEAQQACLDQ DAVIASFDQL YDAWRGGLDW CNAGWLSDGS VQYPITKPRE PCGGQNTVPG VRNYGFWDKD KSRYDVFCFT SNFNGRFYYL IHPTKLTYDE AVQACLNDGA QIAKVGQIFA AWKILGYDRC DAGWLADGSV RYPISRPRRR CSPTEAAVRF VGFPDKKHKL YGVYCFRAYN; Positive Samples: HeLa, A375, Mouse liver; Cellular Location: extracellular matrix, extracellular space.
HAPLN1 belongs to the HAPLN family and contains 1 Ig like V type (immunoglobulin like) domain and 2 Link domains. HAPLN1 stabilizes the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix. Diseases associated with HAPLN1 include Cerebral Creatine Deficiency Syndrome 3 and Rheumatoid Arthritis, Systemic Juvenile.
Specifications
| HAPLN1 | |
| Polyclonal | |
| Unconjugated | |
| HAPLN1 | |
| BB099155; Cartilage link protein; cartilage link protein 1; cartilage linking protein 1; cartilage-link protein; Cartilage-linking protein 1; CLP; CRT1; Crtl1; Crtl1l; fc26f11; fc37g04; fc50e12; fc55h01; HAPLN1; hapln1a; Hyaluronan and proteoglycan link protein 1; hyaluronan and proteoglycan link protein 1a; hyaluronan proteoglycan link protein 1; link protein; LP; LP-1; Proteoglycan link protein; wu:fc26f11; wu:fc37g04; wu:fc50e12; wu:fc55h01 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12950, 1404 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.69 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P10915, Q9QUP5 | |
| HAPLN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 165-354 of human HAPLN1 (NP_0018751). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.