missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAPLN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553925
This item is not returnable.
View return policy
Description
Immunogen sequence: DHLSDNYTLD HDRAIHIQAE NGPHLLVEAE QAKVFSHRGG NVTLPCKFYR DPTAFGSGIH Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.
HAPLN1 belongs to the HAPLN family and contains 1 Ig like V type (immunoglobulin like) domain and 2 Link domains. HAPLN1 stabilizes the aggregates of proteoglycan monomers with hyaluronic acid in the extracellular cartilage matrix. Diseases associated with HAPLN1 include Cerebral Creatine Deficiency Syndrome 3 and Rheumatoid Arthritis, Systemic Juvenile.
Specifications
| HAPLN1 | |
| Polyclonal | |
| Unconjugated | |
| HAPLN1 | |
| BB099155; Cartilage link protein; cartilage link protein 1; cartilage linking protein 1; cartilage-link protein; Cartilage-linking protein 1; CLP; CRT1; Crtl1; Crtl1l; fc26f11; fc37g04; fc50e12; fc55h01; HAPLN1; hapln1a; Hyaluronan and proteoglycan link protein 1; hyaluronan and proteoglycan link protein 1a; hyaluronan proteoglycan link protein 1; link protein; LP; LP-1; Proteoglycan link protein; wu:fc26f11; wu:fc37g04; wu:fc50e12; wu:fc55h01 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1404 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P10915 | |
| HAPLN1 | |
| Recombinant protein corresponding to Human HAPLN1. Recombinant protein control fragment (Product #RP-92474). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction