missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAP40 Recombinant Rabbit Monoclonal Antibody (HL2641)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556733
This item is not returnable.
View return policy
Description
HAP40 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Huntington's disease is caused by an expanded CAG trinucleotide repeat coding for a polyglutamine stretch within the Huntington protein. Huntington co-purifies with a single novel 40 kDa protein designated HAP40. Recombinant HAP40 is cytoplasmic in the presence of Huntington but is actively targeted to the nucleus in the absence of Huntington. These observations suggest that HAP40 °Contributes to the function of normal Huntington and is a candidate for involvement in the aberrant nuclear localization of mutant Huntington found in degenerating neurons in Huntington's disease.
Specifications
| HAP40 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P23610, Q00558 | |
| F8a, F8a1, F8a2, F8a3 | |
| Recombinant fragment of human HAP40. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2641 | |
| Unconjugated | |
| F8a1 | |
| 40-kDa huntingtin-associated protein; AI852759; coagulation factor VIII-associated (intronic transcript) 1; coagulation factor VIII-associated 1; cpG island protein; DXS522E; DXUcsf1; F8A; F8A1; F8A2; F8A3; factor 8-associated gene A; Factor VIII associated protein; factor VIII intron 22 protein; HAP40; huntingtin-associated protein 40; RP23-114O19.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14070, 474383, 474384, 501661, 8263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction