missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAP1 Antibody (1B6), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-74569MFV610
This item is not returnable.
View return policy
Description
HAP1 Monoclonal antibody specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| HAP1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| 1B6 | |
| Flow Cytometry | |
| HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1 | |
| Fusion protein from rat HAP1. [UniProt# P54256] | |
| 0.1 mL | |
| Autophagy, Neuroscience | |
| 9001 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction