missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAO2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | HAO2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18347216
|
Novus Biologicals
NBP3-04697-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392834
|
Novus Biologicals
NBP3-04697-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HAO2 Polyclonal specifically detects HAO2 in Mouse samples. It is validated for Western Blot.Specifications
| HAO2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 51179 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| (S)-2-hydroxy-acid oxidase, peroxisomal, Cell growth-inhibiting gene 16 protein, EC 1.1.3.15, GIG16, growth-inhibiting protein 16, HAOX2glycolate oxidase, hydroxyacid oxidase 2, hydroxyacid oxidase 2 (long chain), Long chain alpha-hydroxy acid oxidase, Long-chain L-2-hydroxy acid oxidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human HAO2 (NP_057611.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title