missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hao2 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 93%.
HAO2 gene ontology annotations related to this gene include fatty acid oxidation.
Specifications
Specifications
| Antigen | Hao2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | HAO2 |
| Gene Accession No. | Q9NYQ2 |
| Gene Alias | (S)-2-hydroxy-acid oxidase, peroxisomal; (S)-hydroxy-acid oxidase, peroxisomal; AI325478; Cell growth-inhibiting gene 16 protein; GIG16; glycolate oxidase; Hao2; Hao-2; Hao3; HAOX2; Haox3; hydroxyacid oxidase (glycolate oxidase) 3; hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); hydroxyacid oxidase 2, kidney; hydroxyacid oxidase 3 (medium-chain); hypothetical protein LOC509481; long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase; medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; medium-chain L-2-hydroxy acid oxidase |
| Gene Symbols | HAO2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?