missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HAO2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15877543
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15877543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877543 Supplier Invitrogen™ Supplier No. PA569135

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 93%; Rat: 100%.

HAO2 gene ontology annotations related to this gene include fatty acid oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAO2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HAO2
Gene Accession No. Q9NYQ3
Gene Alias (S)-2-hydroxy-acid oxidase, peroxisomal; (S)-hydroxy-acid oxidase, peroxisomal; AI325478; Cell growth-inhibiting gene 16 protein; GIG16; glycolate oxidase; Hao2; Hao-2; Hao3; HAOX2; Haox3; hydroxyacid oxidase (glycolate oxidase) 3; hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); hydroxyacid oxidase 2, kidney; hydroxyacid oxidase 3 (medium-chain); hypothetical protein LOC509481; long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase; medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; medium-chain L-2-hydroxy acid oxidase
Gene Symbols HAO2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human HAO2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51179
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.