missing translation for 'onlineSavingsMsg'
Learn More

Hanna™ Instruments Zero Oxygen Solution

Product Code. 11504347
Change view
Click to view available options
For Use With (Equipment):
Polarographic DO Sensors, Galvanic DO Sensor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11504347 Polarographic DO Sensors, Galvanic DO Sensor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504347 Supplier Hanna Instruments Supplier No. HI7040L
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality, bicomponent zero oxygen solutions made from reagent grade chemicals that are contained in hassle-free, easy to mix chemical component bottles.

  • High quality, bicomponent zero oxygen solution made from reagent grade chemicals
  • Prepared with reagent grade chemicals
  • Hassle-free, easy to mix chemical component bottles
  • Air-tight seal to ensure the quality of the solution
  • Bicomponent zero oxygen solution designed for calibrating both polarographic and galvanic dissolved oxygen sensors

Specifications

For Use With (Equipment) Polarographic DO Sensors, Galvanic DO Sensor
compliance-icons compliance-icons
Product Identifier
  • zero oxygen solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sodium metabisulphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.