Learn More
Hanna™ Instruments pH Buffer Solutions
Product Code. 11594307
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
11594307 500 mL 500mL
1 options
Product Code. 11594307 Supplier Hanna Instruments Supplier No. HI7010L
Used for calibration
  • Available in different pH ranges and quantity
  • Calibrated according to NIST standards
  • With/without certificates of analysis
Specifications
pH 10.01
Quantity 500 mL