Learn More
Hanna Instruments™ pH Electrode Cleaning Solution
Product Code. 11584387
Change view
Click to view available options
Capacity (Metric):
230 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
11584387 230 mL Each
1 options
Product Code. 11584387 Supplier Hanna Instruments™ Supplier No. HI7061M
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna Instruments™ pH Electrode Cleaning Solution dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
General purpose cleaning solution that can be used for a variety of applications.
  • Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly.
  • Dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Specifications
Capacity (Metric) 230 mL
Filling Solution Type Aqueous
For Use With (Application) Removing impurities and residues from ph Electrodes