Learn More
DuraLab Hanging Bucket with Clip-On Lid for Laboratory Trolleys
Product Code. 30280259
Change view
Click to view available options
Product Type:
Basket
Unit Size:
Each
Product Code. Product Type Unit Size
30280259 Basket Each
1 options
Product Code. 30280259 Supplier DuraLab Supplier No. DRL0013
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A bucket with a clip-on lid designed to work with the DuraLab Laboratory Trolley range.
  • Attaches to the ends of DuraLab Laboratory Trolleys for convenient placement
  • Clip-on lid keeps contents contained while allowing easy removal
  • Made from durable Polyethylene (PE) material for reliable use
  • Can be used for storing items or collecting waste in laboratory settings
  • Lightweight design allows easy handling and movement
Specifications
Product Type Basket
Color Black
Not compatible with DuraLab Laboratory Trolleys with deep shelves.