Learn More
Brady™ HandiMark™ Indoor/Outdoor Vinyl Tape
Brady's B-595 vinyl will adhere to difficult low surface energy items. The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
Brand:  Brady™ 142279
Code : 50
Additional Details : Weight : 0.11970kg
Product Code. 12830580
  • £93.00 / 1 box
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
  • The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
  • Handimark Consumables Equivalence for BMP71 printers: M71C-500-595-WT (see Related Items).
  • Handimark Consumables Equivalence for BMP61 and M611 printers: M61C-500-595-WT (see Related Items).
Specifications
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Permanent Acrylic, Permanent
Red
Vinyl
Labels
BMP71 (with media adapter), HandiMark
1
1 Roll/Box
-40°F to +180°F
0.004 in.
15.24 m x 25.4 mm
15.24 m
25.4 mm
25.4 mm
HandiMark Indoor/Outdoor Vinyl Tape
Powder
Gloss
Thermal Transfer
Component and equipment labelling, Electrical labelling, Facility Identification, Floor marking, Laboratory labelling, Lean and 5S labelling, Pipe and valve marking, Warehouse marking
60° 50 Gardner Units
-40°C
1 Ea.
-40°C to +82°C
0.102 mm
50 ft.
1 in.
1 in.
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali