Learn More
Particles Plus Handheld Particle Counter 0.3 µm @ 0.1 CFM, Monitors 3 channels simultaneously: 0.3, 0.5, 5.0 µm. Ca
Brand:  Particles Plus 8303
Encompass
Handheld Particle Counter 0.3 µm @ 0.1 CFM, Monitors 3 channels simultaneously: 0.3, 0.5, 5.0 µm. Ca
Product Code. 15624608
  • POA / Each