Learn More
Handheld Electronic Leak Detector Accessories from Restek™
Product Code. 13197112
Encompass_Preferred
Change view
Click to view available options
Product Type:
Small Probe Adapter
Soft Side Carrying Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
13197112 Soft Side Carrying Case
13107122 Small Probe Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Product Code. 13197112 Supplier Restek Supplier No. 22657
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes
Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)
Protects data and analytical columns from costly repairs
  • Optimized sample flow path
  • Sleek ergonomic design
  • Handy probe storage
  • Long-lasting battery: up to 6 hours of continuous use
  • Automatic shut-off capabilities
  • Soft-sided carrying case designed for storage in a smaller space
  • Small probe detector useful to verify hard-to-reach leaks
Certifications
CE marked
Compliance
WEEE, RoHS
Specifications
For Use With (Equipment) Handheld Leak Detector
Product Type Soft Side Carrying Case
Certifications/Compliance CE, WEEE, RoHS
For Use With (Application) Protects data and analytical columns from costly repairs
Includes Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)