Learn More
Handheld Electronic Leak Detector Accessories from Restek™
Product Code. 13107122
Encompass_Preferred
Click to view available options
Product Type:
Small Probe Adapter
Soft Side Carrying Case
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Product Code. 13107122
Brand: Restek 22658
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes
Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)
Protects data and analytical columns from costly repairs
  • Optimized sample flow path
  • Sleek ergonomic design
  • Handy probe storage
  • Long-lasting battery: up to 6 hours of continuous use
  • Automatic shut-off capabilities
  • Soft-sided carrying case designed for storage in a smaller space
  • Small probe detector useful to verify hard-to-reach leaks
Certifications
CE marked
Compliance
WEEE, RoHS
Specifications
For Use With (Equipment) Handheld Leak Detector
Product Type Small Probe Adapter
Certifications/Compliance CE, WEEE, RoHS
For Use With (Application) Protects data and analytical columns from costly repairs
Includes Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)