missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567623
This item is not returnable.
View return policy
Description
HAND1 Polyclonal Antibody for Western Blot, IHC (P)
HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.
Specifications
| HAND1 | |
| Polyclonal | |
| Unconjugated | |
| HAND1 | |
| bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Ehand1; extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; HAND1; heart and neural crest derivatives expressed 1; heart and neural crest derivatives expressed transcript 1; heart- and neural crest derivatives-expressed protein 1; Helix-loop-helix transcription factor expressed in extraembryonic mesoderm and trophoblast; Hxt; Th1; Thing1; thing-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9421 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O96004 | |
| HAND1 | |
| Synthesized peptide derived from human HAND1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction