missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAND1 Antibody (OTI1G10), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71459FR
This item is not returnable.
View return policy
Description
HAND1 Monoclonal specifically detects HAND1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| HAND1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| HAND1 | |
| Full length recombinant protein of human HAND1 (NP_004812) produced in E.coli. | |
| 0.1 mL | |
| 9421.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1G10 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, EHAND, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction