missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hamster PrP (aa 90-102) Synthetic Peptide

Product Code. 11564690
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11564690 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11564690 Supplier Invitrogen™ Supplier No. PEP034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Hamster Prion Protein (residues 90-102) Synthetic Peptide, PEP-034, from Invitrogen.

Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K. PEP-034 is a 13 amino acid synthetic peptide whose sequence corresponds to amino acid residues 90-102 of hamster prion protein. The sequence of this peptide is (amino to carboxy terminus): G (90)-Q-G-G-G-T-H-N-Q-W-N-K-P (102)-G-G-C (Glycine and cysteine residues were added to facilitate coupling KLH).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human prion protein, catalog &Num; PA1-750. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 mL of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 100762792
Species Hamster
Purification Method purified
Common Name Hamster PrP (aa 90-102)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Prnp
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence G(90)-Q-G-G-G-T-H-N-Q-W-N-K-P(102)-G-G-C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.