Learn More
CD49a Hamster anti-Mouse, Brilliant Violet 510, Clone: Ha31/8, BD Optibuild™
Hamster Monoclonal Antibody
Brand: BD Biosciences 740144
Description
The Ha31/8 monoclonal antibody specifically binds to the 180-kDa integrin α 1 chain (CD49a), which is a transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the α 1β 1 (complex known as VLA-1). VLA-1 has been reported to be expressed on activated T cells, monocytes, smooth muscle cells, and endothelial cells. It is a receptor for collagen and laminin. The Ha31/8 monoclonal antibody is specific for both rat and mouse CD49a. It has been reported that Ha31/8 antibody can block VLA-1-mediated binding of rat cells to collagen.
Specifications
| CD49a | |
| Monoclonal | |
| 0.2mg/mL | |
| Itga1; Integrin alpha-1; Integrin α1; Laminin and collagen receptor; VLA-1a | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 λ1 |
| Flow Cytometry | |
| Ha31/8 | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat Emulsified Lewis Rat Glomeruli Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only