Learn More
CD272 Hamster anti-Mouse, Brilliant Violet 421, Clone: HMBT-6B2, BD Optibuild™
Hamster Monoclonal Antibody
Brand: BD Biosciences 740099
Description
The HMBT-6B2 monoclonal antibody specifically binds to CD272 which is also known as B- and T-lymphocyte attenuator (BTLA). CD272 is a type 1 transmembrane glycoprotein and a member of the Ig superfamily. CD272 is variably expressed on developing and mature T and B lymphocytes, NKT cells, NK cells, macrophages and dendritic cells. CD272 expression is upregulated by activated T cells including Th1, Th2, and T follicular helper cells and by anergic T cells. CD272 is structurally similar to CD152/CTLA-4 and CD279/PD-1 and functions as a coinhibitory receptor for B and T cell responses. Herpesvirus entry mediator (HVEM), also known as CD270 and LIGHT-R, has been identified as a ligand for CD272. The crosslinking of CD272 by HVEM inhibits T-cell proliferation and cytokine production.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
| CD272 | |
| Monoclonal | |
| 0.2mg/mL | |
| Btla; B and T lymphocyte attenuator; B- and T-lymphocyte-associated protein | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| HMBT-6B2 | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CD272 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only