Learn More
CD183 Hamster anti-Mouse, Brilliant Violet 650, Clone: CXCR3-173, BD Optibuild™
Hamster Monoclonal Antibody
Brand: BD Biosciences 740630
Description
The CXCR3-173 monoclonal antibody specifically binds to mouse CD183, also known as CXCR3. CD183 is a seven transmembrane spanning, G protein-coupled chemokine receptor for CXC chemokines including CXCL9 (Mig), CXCL10 (IP-10) and CXCL11 (I-TAC). These chemokines are induced by inflammatory cytokines including IFN- γ, IFN- α / β , and TNF. CXCR3 is primarily expressed on activated/memory CD4+ and CD8+ T lymphocytes, Foxp+ regulatory T cells, natural killer T (NKT) cells and mature NK cells. Binding of chemokines to CXCR3 induces integrin activation, cytoskeletal changes, and chemotactic migration of activated lymphocytes. CD183 has been reported to play important roles in T cell recruitment and immune responses in a number of inflammatory and autoimmune diseases. The CXCR3-173 antibody reportedly inhibited in vitro chemotactic responses to CXCL10 or CXCL11 significantly but not to CXCL9. When administered systemically to mouse hosts, the CXCR3-173 antibody reportedly prolonged cardiac and pancreatic islet cell allograft survival. In the presence of CXCR3 ligands, especially, CXCL10 and CXCL11, staining with the antibody can be significantly blocked.
Specifications
| CD183 | |
| Monoclonal | |
| 0.2mg/mL | |
| CXCR3; Cxcr3; CXC-R3; CXCR-3; GPR9; IP-10 Receptor | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| CXCR3-173 | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CXCR3 peptide (amino acids 1-37) | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only