missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAMP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590884
This item is not returnable.
View return policy
Description
Immunogen sequence: MALSSQIWAA CLLLLLLLAS LTSGSVFPQQ TGQLAELQPQ DRAGARASWM PMFQRRRRRD THFPICIFCC GCCHRSKCGM CCKT; Positive Samples: MDA-MB-435.
HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.
Specifications
| HAMP | |
| Polyclonal | |
| Unconjugated | |
| HAMP | |
| HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57817, 84506 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.11 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P81172, Q9EQ21 | |
| HAMP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-84 of human Hepcidin (NP_0669981). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction