missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HAMP Monoclonal Antibody (968018)

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA562033

Product Code. 30534531

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HAMP
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9EQ21
HAMP
Synthetic peptide containing mouse Hepcidin.
100 μg
Primary
Mouse
Antibody
IgG2b
ELISA
968018
Unconjugated
HAMP
HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003
Rat
Protein A/G
RUO
84506
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.