missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PTFE Tubing Adapted to Femaile Luer Lock
Connect to a syringe or other component
Brand: Hamilton™ 90615
Code : NEW
Additional Details : Weight : 0.00376kg
Description
- Made of PTFE
- One female luer fitting on the end
- Tubing is 12 in. in length
Specifications
PTFE | |
Connect to a syringe or other component | |
30.48 cm |
15 G | |
12 in. | |
Tubing Assembly |