missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PTFE Tubing Adapted to Femaile Luer Lock

Connect to a syringe or other component

Brand:  Hamilton™ 90628

Code : Z1

Additional Details : Weight : 2.26800kg

 View more versions of this product

Product Code. 11563094

  • £42.05 / Each
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Made of PTFE
  • One female luer fitting on the end
  • Tubing is 12 in. in length

Specifications

Specifications

PTFE
Connect to a syringe or other component
30.48 cm
28 G
12 in.
Tubing Assembly
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers