missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP™-X200 Cation Exchange Columns
Description
Can be used with any existing HPLC or ion chromatograph to easily determine cations in air, water and soil. PRP-X200 columns provide a number of benefits
Better than Wet Chemical Methods
- Ion chromatography will greatly reduce sample pretreatment and improve the accuracy and precision of results.
- Separate ions at concentrations from 20ppb to 20ppm without suppression or elaborate preconcentration techniques
- Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), a regeneration protocol will usually return your chromatography to its original state.
- Quickly and easily separate five monovalent cations in five minutes or four divalent cations in four minutes
- Highly inert poly(styrene-divinylbenzene) packing allows the use of organic modifier. By varying the concentration of methanol the resolution between cations can be altered.
- One particle size: 10°m
- Four column internal diameters: 2.1 to 10.0mm
- Two column materials: 316 stainless steel and PEEK
- Analytical and semiprep/preparative guard columns
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4.1 mm |
| Flow Rate | 2 mL/min |
| For Use With | Analyzing Inorganic and Organic Cations Using Conductivity or UV Detection |
| Length (Metric) | 150 mm |
| Max. Pressure | 5000 psi (345 bar) |
| Mobile Phase Compatibility | pH 1 to 13, 0 to 100% aqueous, organic modifier |
| Particle Size | 10 μm |
| pH | 1 to 13 |
| Pore Size | 100 Å |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?