Learn More
Hamilton™ PRP™-3 Reversed Phase HPLC Columns
Description
PRP-3 polymeric reversed phase columns, packed with a poly(styrene-divinyl)benzene support, last longer than silica-based columns. Support is pressure stable up to 5,000 psi and crosslinked to prevent shrinkage or swelling when the mobile phase is changed. The support is entirely polymeric and there are no C18 groups to hydrolyze, column lifetime is longer.
Superior pH Stability
- The inherent stability of PRP-3 columns allows their use for any protein purification method from pH 1 to 13. PRP-3 columns allow you to analyze protein purity at low and high pH extremes.
- The 300Å pores on the PRP-3 support help ensure good recovery (>90%) of your protein. The highly inert poly(styrene-divinylbenzene) packing enhances protein recovery because there are no silanol groups on the support to cause irreversible protein adsorption
- Easy to use and ideal for almost any protein separation, including pH conditions from 1 to 13, with organic modifier (0-100%) of virtually any type from routine to the unusual
- A selection of column sizes are available from analytical to semi-prep and preparative scale. Sample scaleup is easy because the PRP-3 packing is consistent from analytical to preparative columns.
- The short analytical (50mm) column is well suited for the gradient elution of high molecular weight proteins. The longer (150mm) column is well suited for smaller proteins.
Product Features:
- Particle size: 10μm and 12-20μL
- Eight column internal diameters: 2.1 to 21.2μL
- Two column materials: 316 stainless steel and PEEK
- Analytical and semiprep/preparative guard columns
- Custom hardware dimensions available
Specifications
Specifications
| Column Format | Preparative Column |
| Diameter (Metric) | 10 mm |
| Length (Metric) | 250 mm |
| Max. Pressure | 5000 psi (345 bar) |
| Particle Size | 10 μm |
| pH | 1 to 13 |
| Pore Size | 300 Å |
| Temperature | 5°C to 85°C |
| USP Type | L21 |
| Stationary Phase | PRP-3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.