Learn More
Hamilton™ Tungsten Cleaning Wires
Product Code. 11588853
Change view
Click to view available options
For Use With (Equipment):
22, 23G and Larger Needle, Syringe and Needle Cleaning Kit
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit
23s Needle, Syringe and Needle Cleaning Kit
24 to 26G Needle, Syringe and Needle Cleaning Kit
26s and 31 to 33G Needle, Syringe and Needle Cleaning Kit
27G Needle, Syringe and Needle Cleaning Kit
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11588853 24 to 26G Needle, Syringe and Needle Cleaning Kit Pack of 10
10464302 23s Needle, Syringe and Needle Cleaning Kit Each
10612503 26s and 31 to 33G Needle, Syringe and Needle Cleaning Kit Pack of 10
10642003 22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit Pack of 10
10345382 27G Needle, Syringe and Needle Cleaning Kit Pack of 10
10427171 22, 23G and Larger Needle, Syringe and Needle Cleaning Kit Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11588853 Supplier Hamilton™ Supplier No. 18303
Only null left
Add to basket
Add to basket
For mechanical cleaning to remove needle obstructions.
Tungsten wires, best used with Hamilton Company Needle Cleaning Concentrate (14-819-85, 14-819-86).
Order Info
Ten wires per pack.
Specifications
Material Tungsten
Length (English) 7 in.
Length (Metric) 17.78 cm
Product Type Cleaning Wire
For Use With (Equipment) 24 to 26G Needle, Syringe and Needle Cleaning Kit