missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Model 7000 (KH) Needle Repair Kit

Eliminates the assembly step which can lead to damage on the smaller syringe volumes. Especially critical for 7000 series syringes which have extremely small plunger wires

Brand:  Hamilton™ 86258

Code : NEW

Additional Details : Weight : 0.04300kg

Product Code. 10271282

  • £274.00 / Each
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For use with 7000 Series syringes only. Needle is 4 in. (10.2cm) in length with Point Style 3. Kit includes knurled hub (KH) needle, ferrule, plunger wire and instructions.

Specifications

Specifications

knurled hub needle, ferrule, plunger wire and instruction, 4 in. L needle with 3 point style
10.2 cm
0.5 μL
4 in.
Repair Kit
7000 Series Syringes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers