Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - Syringes
Product Code. p-4512183
Click to view available options
Includes:
PTFE plunger tip
Plunger tip
Model:
1002.5
1705.5 BFP
Volume (Metric):
1.25 μL
25 μL
Unit Size:
Each
Product Code. 15319742
Brand: Hamilton™ 6608840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose from two syringes designed for use specifically with the Hamilton Microlab 300 Pipetting System
Model 1705.5
  • Bubble Free Prime
  • Expels air from the syringe and valve thanks to an innovative plunger tip
  • 1.25mL
  • Model 1002.5
    • Gastight
    • Ideal for dispensing both liquids and gases
    • Has a precision-machined PTFE plunger tip which creates a leak-free seal
    • 25μL
Specifications
Model 1002.5
Volume (Metric) 1.25 μL
For Use With (Equipment) Microlab™ 300 pipetteting system
Includes PTFE plunger tip